Ac-DL-Pro-OH
Glass bottle for solids, 5 g
Katalognr. BACE4013912.0005
Marketsource
Anbieter: BACHEM BIOCHEMICA
Ac-DL-Pro-OH
Katalognr. BACE4013912.0005
Anbieter: BACHEM BIOCHEMICA
Marketsource
CAS Nummer: 1074-79-9
Direkt zu
Product Details & Documents
Synonyme: 1-Acetylpyrrolidine-2-carboxylic acid, N-Acetyl-DL-Proline
- CAS Nummer:1074-79-9
- Formel:C₇H₁₁NO₃
- MDL-Nummer:MFCD00063230
- Molekulargewicht:157.17 g/mol
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:5 g



