D(-)-Isoleucin
Glass bottle for solids, 5 g
Katalognr. BACE4006515.0005
Marketsource
Anbieter: BACHEM BIOCHEMICA
D(-)-Isoleucin
Katalognr. BACE4006515.0005
Anbieter: BACHEM BIOCHEMICA
Marketsource
CAS Nummer: 319-78-8
Direkt zu
Product Details & Documents
Synonyme: (2R,3R)-(-)-Isoleucine, H-D-Ile-OH
- CAS Nummer:319-78-8
- Formel:H₃CCH₂CH(CH₃)CH(NH₂)COOH
- MDL-Nummer:MFCD00064221
- Molekulargewicht:131.17 g/mol
- Siedepunkt:235 °C (1013 hPa)
- Schmelzender Platingehalt:98 °C
- Dichte:1,224 g/cm³ (20 °C)
- :Density: 1.224 g/cm³ (20 °C)
- EINECS:206-269-2
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



