L(+)-Valin
Glass bottle for solids, 250 g
Katalognr. BACE4031328.0250
Marketsource
Anbieter: BACHEM BIOCHEMICA
Direkt zu
Product Details & Documents
Synonyme: H-Val-OH, Valine, (S)-(+)-Valine
- CAS Nummer:72-18-4
- Formel:C₅H₁₁NO₂
- MDL-Nummer:MFCD00064220
- Molekulargewicht:117.15 g/mol
- Merck-Index:13,09458
- Siedepunkt:217 °C (1013 hPa)
- Schmelzender Platingehalt:86 °C
- Dichte:1,284 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:200-773-6
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:250 g



