L-Threonin
Glasflasche für Feststoffe, 1 kg
Katalognr. BACE4031298.1000
Marketsource
Anbieter: BACHEM BIOCHEMICA
Direkt zu
Product Details & Documents
Synonyme: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
- CAS Nummer:72-19-5
- Formel:C₄H₉NO₃
- MDL-Nummer:MFCD00064270
- Molekulargewicht:119.12 g/mol
- Lagertemperatur:Raumtemperatur
- EINECS:200-774-1
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



