Containers with screw cap
200 ml, Without label, Individually wrapped, PS
Catalog # ANICC200S120
Supplier: Anicrin
Jump to:
Product Details & Documents
Containers made of PS or PP with colour coded screw caps.
- Good chemical resistance
- Good temperature resistance
Ordering information: Specify the cap colour (red, yellow and white) required.
Caution: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Documents
ANICC200S110_TDS_EN
Datasheet
Specifications
- Verpackt:Individually wrapped
- Ausführung:Without label
- Kapazität:200 ml
- Material:PS
- Seq:03