Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Natriumhydroxid ≥97%, Plätzchen ACS

Natriumhydroxid ≥97%, Plätzchen ACS

Pail, 5 kg
Catalog # 0583-5KG
Corrosive
Supplier:   VWR Chemicals
Natriumhydroxid ≥97%, Plätzchen ACS
Catalog # 0583-5KG
Supplier:  VWR Chemicals
CAS Number:  1310-73-2
Natriumhydroxid ≥97%, Plätzchen ACS
Natriumhydroxid ≥97%, Plätzchen ACS

Pail, 5 kg

CAS Number:  1310-73-2
Price
SFr. 528.00
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2,13 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specification Test Results

  • Calcium:
    < 0,005%
  • Chloride:
    ≤ 0,005%
  • Schwermetalle:
    ≤ 0,002%
  • Iron:
    ≤ 0,001%
  • Magnesium:
    ≤ 0,002%
  • Mercury:
    ≤ 0,1 ppm
  • Nickel:
    ≤ 0,001%
  • Stickstoff-Bestandteile:
    ≤ 0,001%
  • Phosphate:
    ≤ 0,001%
  • Potassium:
    ≤ 0,02%
  • Purity:
    ≥ 97%
  • Natriumcarbonat:
    ≤ 1%
  • Sulphate:
    ≤ 0,003%

Specifications

  • Packung:
    Pail
  • VE:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.