Iris scissors
Scharf/scharf, 115 mm
Catalog # MART11-982-11
Supplier: KLS Martin Group
Jump to:
Product Details & Documents
Dissecting scissors made of stainless steel. One model features a TC Gold finish.
- TC Gold finish: Tungsten Carbide inlays provide an exceptionally long service life
- Straight or curved available
Specifications
- Typ:Scharf/scharf
- Länge:115 mm
- Seq:02