Trypsinizing flasks, WHEATON®
300 ml, Schraubverschluss oben, 38-430, Schraubverschluss, 20-415
Katalognr. WHEA355755
Anbieter: DWK Life Sciences
Direkt zu
Product Details & Documents
Borosilicate glass, clear.
- With pour out and caps
- Fluted design improves mixing
- Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
Technische Daten
- Kapazität:300 ml
- Bezeichnung:Schraubverschluss oben, 38-430
- Hals:Schraubverschluss, 20-415