AuftragsverfolgungKontakt
Trypsinising flasks, with screw caps, WHEATON® | Avantor
Trypsinising flasks, with screw caps, WHEATON®
35 ml, Schraubverschluss oben, 24-430
Katalognr. WHEA355442
Anbieter:   DWK Life Sciences
Trypsinising flasks, with screw caps, WHEATON®
Katalognr. WHEA355442
Anbieter:  DWK Life Sciences
Trypsinising flasks, with screw caps, WHEATON®
Trypsinising flasks, with screw caps, WHEATON®
35 ml, Schraubverschluss oben, 24-430
Preis
SFr. 133.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
Zertifizierung: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
Technische Daten
  • Kapazität:
    35 ml
  • Bezeichnung:
    Schraubverschluss oben, 24-430
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.