Caps for bottles, open top, Microlink®, WHEATON®
Screw cap, solid top, with PP/silicone/PTFE liner, 24-400, Weiß
Katalognr. WHEA240251-01
Anbieter: DWK Life Sciences
Caps for bottles, open top, Microlink®, WHEATON®
Katalognr. WHEA240251-01
Anbieter: DWK Life Sciences
Direkt zu
Product Details & Documents
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
- Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
Technische Daten
- Gewinde:24-400
- Farbe:Weiß
- Bezeichnung:Screw cap, solid top, with PP/silicone/PTFE liner
- Seq:05