Caps for bottles, open top, Microlink®, WHEATON®
Schraubverschluss, Polypropylen/Silikon/PTFE-Einlage, oben geschlossen, 20-400, Weiß
Katalognr. WHEA240250-01
Anbieter: DWK Life Sciences
Caps for bottles, open top, Microlink®, WHEATON®
Katalognr. WHEA240250-01
Anbieter: DWK Life Sciences
Direkt zu
Product Details & Documents
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
- Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
Technische Daten
- Gewinde:20-400
- Farbe:Weiß
- Bezeichnung:Schraubverschluss, Polypropylen/Silikon/PTFE-Einlage, oben geschlossen
- Seq:02