Trimming knives
SM 92
Katalognr. SWAN4202
Anbieter: SWANN MORTON
Direkt zu
Product Details & Documents
Heavy duty trimming knifes with fixed or retractable blade.
- Blades can be exchanged
- Trimming knife with a smart black holster that clips onto a belt or waistband
Lieferinformationen: The fixed trimming knife is supplied with holster and three SM92 industrial blades. The retractable knife is supplied with two SM92 industrial blades.
Technische Daten
- Typ:SM 92
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.