AuftragsverfolgungKontakt
Trimming knives | Avantor
Trimming knives
SM 92
Katalognr. SWAN4202
Anbieter:   SWANN MORTON
Trimming knives
Katalognr. SWAN4202
Anbieter:  SWANN MORTON
Trimming knives
Trimming knives
SM 92
Preis
42.60 SFr.
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents
Heavy duty trimming knifes with fixed or retractable blade.
  • Blades can be exchanged
  • Trimming knife with a smart black holster that clips onto a belt or waistband
Lieferinformationen: The fixed trimming knife is supplied with holster and three SM92 industrial blades. The retractable knife is supplied with two SM92 industrial blades.
Technische Daten
  • Typ:
    SM 92