Trimming knives
SM 96
Katalognr. SWAN4203
Anbieter: SWANN MORTON
Direkt zu
Product Details & Documents
Heavy duty trimming knifes with fixed or retractable blade.
- Blades can be exchanged
- Trimming knife with a smart black holster that clips onto a belt or waistband
: The fixed trimming knife is supplied with holster and three SM92 industrial blades. The retractable knife is supplied with two SM92 industrial blades.
Technische Daten
- Typ:SM 96