Ammoniumnitrat
5 kg
Catalog # EURLEMR281005
Supplier: EUROCLONE
Jump to:
Product Details & Documents
- CAS Number:6484-52-2
- Formula:NH₄NO₃
- MDL Number:MFCD00011425
- Molecular Weight:80.04 g/mol
- Merck Index:13,00538
- Boiling Pt:230 °C (1013 hPa)
- Melting Pt:169 °C
- Density:1,73 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:229-347-8
- UN:1942
- ADR:5.1,III
Specifications
Specifications
- VE:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



