Kaliumnitrat
Glass bottle for solids, 1 kg
EURLEMR280001
: Nitrate of Potash
- :7757-79-1
- :KNO₃
- :MFCD00011409
- :101.1 g/mol
- :13,07733
- :334 °C
- :2,109 g/cm³ (20 °C)
- :Raumtemperatur
- :231-818-8
- :1486
- :5.1,III
- :01-2119488224-35
- Packung:Glass bottle for solids
- VE:1 kg