Silbernitrat
1 KIT
EURLEMR267100
: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- :7761-88-8
- :AgNO₃
- :MFCD00003414
- :169.87 g/mol
- :12,08661
- :444 °C (1013 hPa)
- :210 °C
- :4,35 g/cm³ (20 °C)
- :Raumtemperatur
- :231-853-9
- :1493
- :5.1,II
- :01-2119513705-43
- VE:1 KIT