Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Silbernitrat

Silbernitrat

50 g
Catalog # EURLEMR267050
Oxidizing
Corrosive
Environment
Supplier:   EUROCLONE
Silbernitrat
Catalog # EURLEMR267050
Supplier:  EUROCLONE
CAS Number:  7761-88-8
Silbernitrat
Silbernitrat

50 g

CAS Number:  7761-88-8
Price
Inquire For Price
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:210 °C
  • Density:4,35 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Specifications

Specifications

  • VE:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.