Tween® 20 (Polysorbat)
1 KIT
Catalog # EURLEMR243005
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monolaurate
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Molecular Weight:1227.53 g/mol
- Density:1,1 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:500-018-3
Specifications
Specifications
- VE:1 KIT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



