Trypanblau
1 KIT
Catalog # EURLEMR241100
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
- CAS Number:72-57-1
- Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
- MDL Number:MFCD00003969
- Molecular Weight:960.82 g/mol
- EINECS:200-786-7
Specifications
Specifications
- VE:1 KIT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



