Natriumpyruvat
Glasflasche für Feststoffe, 1 kg
Catalog # EURLEMR213001
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Number:113-24-6
- Formula:CH₃COCOONa
- MDL Number:MFCD00002586
- Molecular Weight:110.04 g/mol
- Melting Pt:220…230 °C
- Density:1,718 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:204-024-4
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:1 kg



