Ribonuclease A
Glass bottle for solids, 100 mg
Katalognr. EURLEMR199100
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:100 mg
- Cat. No.:EURLEMR199100
- CAS Nr.:9001-99-4
- Enzym-Synonyme:RNase A
- Enzymname:Ribonuclease A
- MDL No.:MFCD00132184
- Lagerbedingungen:Freezer



