Ribonuclease A
Glass bottle for solids, 1 g
Katalognr. EURLEMR199001
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 g
- Cat. No.:EURLEMR199001
- CAS Nr.:9001-99-4
- Enzym-Synonyme:RNase A
- Enzymname:Ribonuclease A
- MDL No.:MFCD00132184
- Lagerbedingungen:Freezer
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



