Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Methanol

Methanol

1 KIT
Catalog # EURLEMR173025
Flammable
Toxic
Health
Supplier:   EUROCLONE
Methanol
Catalog # EURLEMR173025
Supplier:  EUROCLONE
CAS Number:  67-56-1
Methanol
Methanol

1 KIT

CAS Number:  67-56-1
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64,6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0,7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (geschlossener Tiegel)
  • Storage Temperature:Raumtemperatur
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specifications

  • VE:
    1 KIT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.