Formamid
Glasflasche, 250 mL
Katalognr. EURLEMR141250
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Synonyme: Carbamaldehyde
- CAS Nummer:75-12-7
- Formel:HCONH₂
- MDL-Nummer:MFCD00007941
- Molekulargewicht:45.04 g/mol
- Merck-Index:13,04261
- Siedepunkt:210 °C (1013 hPa)
- Schmelzender Platingehalt:2 °C
- Dichte:1,1334 g/cm³ (20 °C)
- Blitz Pt:79 °C
- Lagertemperatur:Kühlschrank
- EINECS:200-842-0
Technische Daten
Technische Daten
- Packung:Glasflasche
- VE:250 mL
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



