D(+)-Saccharose
1 KIT
EURLEMR119500
: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
- :57-50-1
- :C₁₂H₂₂O₁₁
- :MFCD00006626
- :342.3 g/mol
- :13,08966
- :169…170 °C
- :1,29 g/cm³ (20 °C)
- :Raumtemperatur
- :200-334-9
- VE:1 KIT