D(+)-Maltose Monohydrat
Glass bottle for solids, 250 g
EURLEMR117250
: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
- :6363-53-7
- :C₁₂H₂₂O₁₁·1H₂O
- :MFCD00149343
- :360.32 g/mol
- :13,05736
- :160…165 °C
- :Raumtemperatur
- :200-716-5
- Packung:Glass bottle for solids
- VE:250 g