D(+)-Biotin
Glass bottle for solids, 250 mg
EURLEMR115250
: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- :58-85-5
- :C₁₀H₁₆N₂O₃S
- :MFCD00005541
- :244.31 g/mol
- :13,01231
- :232…233 °C
- :1,41 g/cm³ (20 °C)
- :Kühlschrank
- :200-399-3
- Packung:Glass bottle for solids
- VE:250 mg