D(+)-Biotin
1 KIT
Katalognr. EURLEMR115001
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Synonyme: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Nummer:58-85-5
- Formel:C₁₀H₁₆N₂O₃S
- MDL-Nummer:MFCD00005541
- Molekulargewicht:244.31 g/mol
- Merck-Index:13,01231
- Schmelzender Platingehalt:232…233 °C
- Dichte:1,41 g/cm³ (20 °C)
- Lagertemperatur:Kühlschrank
- EINECS:200-399-3
Technische Daten
Technische Daten
- VE:1 KIT
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



