Brij® 35
Glasflasche für Feststoffe, 250 g
Katalognr. EURLEMR101250
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Synonyme: Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
- CAS Nummer:9002-92-0
- Formel:C₅₈H₁₁₈O₂₄
- MDL-Nummer:MFCD00080891
- Molekulargewicht:1199.56 g/mol
- Lagertemperatur:Raumtemperatur
- EINECS:500-002-6
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



