Bisacrylamid
1 KIT
Katalognr. EURLEMR095050
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Synonyme: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
- CAS Nummer:110-26-9
- Formel:(H₂C=CHCONH)₂CH₂
- MDL-Nummer:MFCD00008625
- Molekulargewicht:154.17 g/mol
- Siedepunkt:125 °C (3,33 hPa)
- Schmelzender Platingehalt:185 °C
- Dichte:1,235 g/cm³ (20 °C)
- Blitz Pt:138 °C
- Lagertemperatur:Kühlschrank
- EINECS:203-750-9
Technische Daten
Technische Daten
- VE:1 KIT
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



