Ammoniumsulfat
Glass bottle for solids, 1 kg
EURLEMR092001
: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
- :7783-20-2
- :(NH₄)₂SO₄
- :MFCD00003391
- :132.14 g/mol
- :13,00559
- :336…339 °C
- :1,78 g/cm³ (20 °C)
- :Raumtemperatur
- :231-984-1
- :01-2119455044-46
- Packung:Glass bottle for solids
- VE:1 kg