Ammoniumchlorid
5 kg
Katalognr. EURLEMR089005
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Synonyme: Ammonium muriate, Salmiac
- CAS Nummer:12125-02-9
- Formel:NH₄Cl
- MDL-Nummer:MFCD00011420
- Molekulargewicht:53.49 g/mol
- Merck-Index:13,00510
- Schmelzender Platingehalt:335 °C
- Dichte:1,5256 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:235-186-4
Technische Daten
Technische Daten
- VE:5 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



