Agar
1 KIT
Catalog # EURLEMR083100
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- Storage Temperature:Raumtemperatur
- EINECS:232-658-1
Specifications
Specifications
- VE:1 KIT



