Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Aceton
Aceton
1 KIT
  EURLEMR081025
 :   EUROCLONE
Aceton
  EURLEMR081025
 :  EUROCLONE
 :  67-64-1
Aceton
Aceton
1 KIT
 :  67-64-1
1 * 1 KIT
 :  2-Propanone, DMK, Propanone, Dimethyl ketone
  •  :67-64-1
  •  :CH₃COCH₃
  •  :MFCD00008765
  •  :58.08 g/mol
  •  :13,00067
  •  :56,2 °C (1013 hPa)
  •  :–95,4 °C
  •  :0,792 g/cm³ (20 °C)
  •  :<–20 °C (geschlossener Tiegel)
  •  :Raumtemperatur
  •  :200-662-2
  •  :1090
  •  :3,II
  •  :01-2119471330-49
  • VE:
    1 KIT
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.