Aceton
1 KIT
Katalognr. EURLEMR081001
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Synonyme: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Nummer:67-64-1
- Formel:CH₃COCH₃
- MDL-Nummer:MFCD00008765
- Molekulargewicht:58.08 g/mol
- Merck-Index:13,00067
- Siedepunkt:56,2 °C (1013 hPa)
- Schmelzender Platingehalt:–95,4 °C
- Dichte:0,792 g/cm³ (20 °C)
- Blitz Pt:<–20 °C (geschlossener Tiegel)
- Lagertemperatur:Raumtemperatur
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- ERREICHEN:01-2119471330-49
Technische Daten
Technische Daten
- VE:1 KIT
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



