Deoxythymidine triphosphate (dTTP)
Glasflasche für Feststoffe, 25 mg
Catalog # EURLEMR073025
Supplier: EUROCLONE
Deoxythymidine triphosphate (dTTP)
Catalog # EURLEMR073025
Supplier: EUROCLONE
CAS Number: 18423-43-3
Jump to:
Product Details & Documents
Synonyms: Thymidine-5'-triphosphoric acid, Deoxythymidine triphosphate, TPP, 2'-Deoxythymidine-5'-triphosphoric acid, dTTP
- CAS Number:18423-43-3
- Formula:C₁₀H₁₇N₂O₁₃P₃
- MDL Number:MFCD00080338
- Molecular Weight:466.17 g/mol
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:25 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



