Polyethylenglycol 8.000
Glasflasche für Feststoffe, 1 kg
Katalognr. EURLEMR037001
Anbieter: EUROCLONE
Direkt zu
Product Details & Documents
Synonyme: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
- CAS Nummer:25322-68-3
- Formel:H(OCH₂CH₂)nOH
- MDL-Nummer:MFCD01779615
- Molekulargewicht:8000 g/mol
- Merck-Index:12,07729
- Siedepunkt:250 °C (1013 hPa)
- Dichte:1,125 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:500-038-2
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



