Calciumchlorid
1 KIT
Catalog # EURLEMR026500
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
- CAS Number:10043-52-4
- Formula:CaCl₂
- MDL Number:MFCD00010903
- Molecular Weight:110.98 g/mol
- Merck Index:14,01659
- Boiling Pt:1600 °C (1013 hPa)
- Melting Pt:772 °C
- Density:2,174 g/cm³ (20 °C)
- Storage Temperature:Raumtemperatur
- EINECS:233-140-8
- REACH:01-2119494219-28
Specifications
Specifications
- VE:1 KIT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



