Gouges, Aesculap®
Gouge for taking grafts fro the ribs, Walter, 10 mm, 205 mm
Katalognr. AESCFL583R
Anbieter: Aesculap
Direkt zu
Product Details & Documents
Technische Daten
- Typ:Walter
- Breite:10 mm
- Bezeichnung:Gouge for taking grafts fro the ribs
- Länge:205 mm
- Seq:41