Micro scissors for endoscopic work, Aesculap® MINOP®
FF386R, blunt/blunt, 265 mm
Katalognr. AESCFF386R
Anbieter: Aesculap
Direkt zu
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
- Very delicate
Ø = 2 mm
Ordering information: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Technische Daten
- Typ:FF386R, blunt/blunt
- Länge:265 mm
- Seq:03