Dilator set, Aesculap®
Dilator, 5 mm
Katalognr. AESCEM105R
Anbieter: Aesculap
Direkt zu
Product Details & Documents
Complete set of uterine dilators.
Delivery information: Consists of 26 dilators, 1 rack for dilators (1 to 17 mm), 1 rack for dilators (18 to 26 mm) and metal case with hinged aluminium lid (330×220×80 mm).
Technische Daten
- Bezeichnung:Dilator, 5 mm
- Seq:06