Anoscopes, Bensaude, Aesculap®
Anoscope, Bensaude, 55 mm, 17 mm
Katalognr. AESCEA862C
Anbieter: Aesculap
Direkt zu
Product Details & Documents
Anoscopes for abdominal surgery.
Technische Daten
- Ø:17 mm
- Bezeichnung:Anoscope, Bensaude
- Länge:55 mm
- Seq:01