Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Chisels, Aesculap® | Avantor

Chisels, Aesculap®

Chisel, curved, with PEEK handle, -, 200 mm, 20 mm
  AESCDO755R
 :   Aesculap
Chisels, Aesculap®
  AESCDO755R
 :  Aesculap
Chisels, Aesculap®

Chisel, curved, with PEEK handle, -, 200 mm, 20 mm

 
 
   
1 * 1 ST
 

These chisels are used for dental surgery.

 
  • Bezeichnung:
    Chisel, curved, with PEEK handle
  • Länge:
    200 mm
  • Seq:
    28
  • Typ:
    -
  • Breite:
    20 mm