Chisels, Aesculap®
Chisel, Lucas, 160 mm, 6 mm
Katalognr. AESCDO713R
Anbieter: Aesculap
Direkt zu
Product Details & Documents
These chisels are used for dental surgery.
Technische Daten
- Bezeichnung:Chisel
- Länge:160 mm
- Typ:Lucas
- Breite:6 mm
These chisels are used for dental surgery.