Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Forceps, micro, jeweller pattern, Aesculap® | Avantor

Forceps, micro, jeweller pattern, Aesculap®

Curved tip, very fine (0,2 mm), 115 mm
Katalognr. AESCBD333R
Anbieter:   Aesculap
Forceps, micro, jeweller pattern, Aesculap®
Katalognr. AESCBD333R
Anbieter:  Aesculap
Forceps, micro, jeweller pattern, Aesculap®

Curved tip, very fine (0,2 mm), 115 mm

Preis
47.80 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Technische Daten
  • Typ:
    Curved tip, very fine (0,2 mm)
  • Länge:
    115 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.