Dissecting forceps, Overholt type, Aesculap®
Fine pattern, figure 6, 300 mm
Katalognr. AESCBJ096R
Anbieter: Aesculap
Direkt zu
Product Details & Documents
Curved dissecting and ligature forceps. Different figures available.
Technische Daten
- Typ:Fine pattern, figure 6
- Länge:300 mm
- Seq:10