Dissecting forceps, Overholt type, Aesculap®
Fine pattern, figure 6, 225 mm
Catalog # AESCBJ086R
Supplier: Aesculap
Jump to:
Product Details & Documents
Curved dissecting and ligature forceps. Different figures available.
Specifications
- Typ:Fine pattern, figure 6
- Länge:225 mm
- Seq:09
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.