Dosen mit Deckel
50 mm, 50 mm
Katalognr. BOCH8770
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Dressing jar either with flat or high shape, made of aluminium.
- Very good temperature resistance
- Good chemical resistance
- Lightweight
Technische Daten
- Ext.Ø:50 mm
- Höhe:50 mm
- Seq:04