Evaporating dishes, high shape
2000 ml, 105 mm, 160 mm
BOCH8564
: BOCHEM
Evaporating dish, stainless steel, with contact bottom for heating plates and magnetic stirrers.
- Very good temperature resistance
- Good chemical resistance
- Easy to clean
- Ext.Ø:160 mm
- Kapazität:2000 ml
- Höhe:105 mm