Support clips
Shaft, M12, for support clips, length 300 mm, O. Ø 15 mm
Katalognr. BOCH582
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
These support clips are coated with ceramic inside.
- For tubes and cylindrical vessels
- With fitting
Bestellinformationen: The shafts (rods) need to be ordered separately.
Technische Daten
- Bezeichnung:Shaft, M12, for support clips, length 300 mm, O. Ø 15 mm